2-Bromo-6-nitrobenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR55195-5G OR55195-10G OR55195-1G
76592-332EA 105 USD
76592-332 76592-330 76592-328
2-Bromo-6-nitrobenzoic acid
2-Bromo-6-nitrobenzoic acid
Formula: C₇H₄BrNO₄
MW: 246.0200 g/mol
Melting Pt: 177…178 °C
Storage Temperature: Ambient
MDL Number: MFCD13688997
CAS Number: 38876-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR