(R)-(-)-Prolinol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: D-ProlinolD(-)-Prolinol(2R)-pyrrolidin-2-ylmethanol

41831-0050 41831-0010
TS41831-0010EA 71 USD
TS41831-0010 TS41831-0050
(R)-(-)-Prolinol 99%
(R)-(-)-Prolinol
Formula: C₅H₁₁NO
MW: 101.1 g/mol
Boiling Pt: 75 °C (2 mmHg)
Density: 1.02 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00064321
CAS Number: 68832-13-3

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4830 to 1.4870 (20 °C, 589 nm)
GC ≥98.5 %
Specific optical rotation -29° to -33° (20 °C, 589 nm) (c=1, toluene)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR