Imidazole-4,5-dicarboxylic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,5-Imidazoledicarboxylic acid1H-Imidazole-4,5-dicarboxylic acid

41225-0250
TS41225-0250EA 143 USD
TS41225-0250
Imidazole-4,5-dicarboxylic acid 99%
Imidazole-4,5-dicarboxylic acid
Formula: C₅H₄N₂O₄
MW: 156.10 g/mol
Melting Pt: 265 °C
Storage Temperature: Ambient
MDL Number: MFCD00005200
CAS Number: 570-22-9
EINECS: 209-327-5

Order Now

Specification Test Results

Appearance Light yellow to yellow-brown Crystalline powder
Infrared spectrum Conforms
Titration excess NaOH ≥98.5 %
UV at 233 nm A: ≤1
UV at 240 nm A: ≤0.2
UV at 250 nm A: ≤0.02
UV at 260 nm A: ≤0.005
UV at 270 nm A: ≤0.005
UV at 280 nm A: ≤0.005
UV 1 cm cell vs HPLC-grade water
Color scale ≤5 APHA
Stabilizer Approx. 0.4 % Methanol
Residue after evaporation ≤0.0002 %
Acidity ≤0.0001 meq/g
Filtered to 0.2 micrometer Filtered
Free chlorine ≤0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR