(±)-Cbz-α-phosphonoglycine trimethyl ester 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-{[(benzyloxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetateMethyl 2-(((benzyloxy)carbonyl)amino)-2-(dimethoxyphosphoryl)acetate(±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate(±)-Benzyloxycarbonyl-α-phosphonoglycine trimethyl ester

37004-0050
TS37004-0050EA 155.04 USD
TS37004-0050
(±)-Cbz-α-phosphonoglycine trimethyl ester 95%
(±)-Cbz-α-phosphonoglycine trimethyl ester
Formula: C₁₃H₁₈NO₇P
MW: 331.27 g/mol
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00043304
CAS Number: 88568-95-0

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 77 to 80 °C
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR