1H-Benzimidazole-5-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Benzo[d]imidazole-5-carboxylic acidBenzimidazole-5-carboxylic acid

29087-0050 29087-0250
TS29087-0250EA 161 USD
TS29087-0250 TS29087-0050
1H-Benzimidazole-5-carboxylic acid 98%
1H-Benzimidazole-5-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00011555
CAS Number: 15788-16-6

Order Now

Specification Test Results

Appearance Grey to brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR