D(+)-α-Acetobromoglucose 98.5%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bromo-2,3,4,6-Tetra-o-Acetyl-α-D-Glucopyranose2,3,4,6-Tetra-O-acetyl-α-D-glucopyranosyl bromideAcetobromo glucoseAcetobromo-α-D-glucose

27093-0500 27093-0100
TS27093-0500EA 214.2 USD
TS27093-0500 TS27093-0100
D(+)-α-Acetobromoglucose 98.5%
D(+)-α-Acetobromoglucose
Formula: C₁₄H₁₉BrO₉
MW: 411.2 g/mol
Melting Pt: 83…88 °C
Storage Temperature: Freezer
MDL Number: MFCD00063254
EINECS: 209-339-0

Order Now

Specification Test Results

Appearance White to beige to light grey Powder
Infrared spectrum Conforms
Melting point 84 to 89 °C
Titration Argentometric ≥98.0 %
Specific optical rotation +188° to +202° (20 °C, 589 nm) (c=2, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR