Thionin(acetate), pure, high purity biological stain

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,7-Diaminophenothiazin-5-ium acetate

22984-0250 22984-0050
TS22984-0050EA 67.32 USD
TS22984-0050 TS22984-0250
Thionin(acetate), pure, high purity biological stain
Thionin (Acetate)
Formula: C₁₄H₁₃N₃O₂S
MW: 287.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00081194
CAS Number: 78338-22-4

Order Now

Specification Test Results

Appearance Dark green Crystalline powder or crystals
Infrared spectrum Conforms
UV spectrum Complies
Dye content ≥85 %
UV Lambda max 597 to 601 nm (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR