Creatinine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one

22894-2500 22894-0250 22894-0500
TS22894-0250EA 53.55 USD
TS22894-0250 TS22894-2500 TS22894-0500
Creatinine 98%
Creatinine
Formula: C₄H₇N₃O
MW: 113.12 g/mol
Melting Pt: 255 °C (dec.)
Flash Pt: 290 °C
Storage Temperature: Ambient
MDL Number: MFCD00059730
CAS Number: 60-27-5
EINECS: 200-466-7

Order Now

Specification Test Results

Appearance White to yellowish-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (105 °C)
Sulfated ash ≤0.2 %
Solubility (5 % in water) Clear colorless to slightly yellow
Titration with HClO4 97.5 to 102.5 % (on dried substance)
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR