Cacodylic acid sodium salt trihydrate 98%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dimethylarsonic acid sodium salt trihydrate
, Sodium cacodylate trihydrate
, Sodium dimethylarsinate trihydrate
, Dimethylarsinic acid sodium salt trihydrate
21497-0100
21497-1000
21497-5000
21497-0250
TS21497-1000EA
259
USD
TS21497-1000
TS21497-0100
TS21497-5000
TS21497-0250
Cacodylic acid sodium salt trihydrate 98%, pure
Cacodylic acid sodium salt trihydrate
Formula:
(CH₃)₂As(O)ONa·3H₂O MW: 213.94 g/mol Melting Pt: 77…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149079 CAS Number: 6131-99-3 EINECS: 204-708-2 UN: 1688 ADR: 6.1,II |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration with HCl | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...