Thiophene-2-methylamine 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Thiophenemethylamine2-(Aminomethyl)thiophene

20557-0250 20557-1000
TS20557-0250EA 134.64 USD
TS20557-0250 TS20557-1000
Thiophene-2-methylamine 95%
Thiophene-2-methylamine
Formula: C₅H₇NS
MW: 113.19 g/mol
Boiling Pt: 95…99 °C (28 mmHg)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00005460
CAS Number: 27757-85-3
EINECS: 248-639-6
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR