4-Bromophenethylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(4-Bromophenyl)ethylamine

18309-0050 18309-0010
TS18309-0050EA 143 USD
TS18309-0050 TS18309-0010
4-Bromophenethylamine 98%
4-Bromophenethylamine
Formula: C₈H₁₀BrN
Boiling Pt: 63…72 °C (.2 mmHg)
Density: 1.29 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008189
EINECS: 277-636-2
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5730 to 1.5750 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR