3,5-Dimethylpiperidin (mixture of cis- and trans- isomers) 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,5-Dimethylpiperidine

18123-1000 18123-0250
TS18123-1000EA 182.58 USD
TS18123-1000 TS18123-0250
3,5-Dimethylpiperidin (mixture of cis- and trans- isomers) 96%
3,5-Dimethylpiperidin (mixture of cis- and trans-isomers)
Formula: C₇H₁₅N
MW: 113.20 g/mol
Boiling Pt: 144 °C
Density: 0.85 g/cm³
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00005996
CAS Number: 35794-11-7
EINECS: 252-730-6
UN: 2733
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4434 to 1.4464 (20 °C, 589 nm)
Water ≤0.5 %
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR