Methyl-α-D-glucopyranoside 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triolα-D-Methylglucoside

17384-0025 17384-1000 17384-5000
TS17384-5000EA 104.04 USD
TS17384-5000 TS17384-1000 TS17384-0025
Methyl-α-D-glucopyranoside 99%
Methyl α-D-glucopyranoside
Formula: C₇H₁₄O₆
MW: 194.18 g/mol
Boiling Pt: 200 °C (.2 mmHg)
Melting Pt: 169…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00064086
EINECS: 202-571-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.1 % (60 to 70 °C)
HPLC ≥98.5 % (ELSD)
Specific optical rotation +156° to +161° (20 °C, 589 nm) (c=10, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR