Hydantoin 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Imidazolidine-2,4-dione

16870-1000 16870-5000
TS16870-5000EA 147.9 USD
TS16870-5000 TS16870-1000
Hydantoin 99%
Hydantoin
Formula: C₃H₄N₂O₂
MW: 100.08 g/mol
Melting Pt: 220…223 °C
Storage Temperature: Ambient
MDL Number: MFCD00005259
CAS Number: 461-72-3
EINECS: 207-313-3

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 220 to 223 °C
Titration with NaOH 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR