Strychnine 98%

Supplier: Thermo Scientific Chemicals
Danger

15895-1000 15895-0250
TS15895-0250EA 104.04 USD
TS15895-0250 TS15895-1000
Strychnine 98%
Strychnine
Formula: C₂₁H₂₂N₂O₂
MW: 334.42 g/mol
Melting Pt: 284…286 °C
Storage Temperature: Ambient
MDL Number: MFCD00005941
CAS Number: 57-24-9
EINECS: 200-319-7
UN: 1692
ADR: 6.1,I

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %
Chloride (Cl) ≤0.01 %
Iron (Fe) ≤0.001 %
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR