4-Methylphenethylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(p-Tolyl)ethylamine

15788-0010 15788-0050
TS15788-0050EA 156 USD
TS15788-0050 TS15788-0010
4-Methylphenethylamine 97%
4-Methylphenethylamine
Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 214 °C (760 mmHg)
Density: 0.93 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00008195
CAS Number: 3261-62-9
EINECS: 221-865-2
UN: 2922
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5280 (20 °C, 589 nm)
GC ≥96.0 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR