Methyl-3-aminocrotonate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-aminobut-2-enoate

15679-1000 15679-5000 15679-0050
TS15679-1000EA 34.07 USD
TS15679-1000 TS15679-5000 TS15679-0050
Methyl-3-aminocrotonate 97%
Methyl 3-aminocrotonate
Formula: C₅H₉NO₂
MW: 115.13 g/mol
Melting Pt: 81…83 °C
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00008072
EINECS: 238-056-5

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 81 to 85 °C
Titration with HClO4 ≥96.0 %
Sulfate (SO4) ≤0.005 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤5 ppm
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %
Ammonia (NH3) ≤0.003 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR