Saccharin 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
o-Sulphobenzimide
, o-Benzoic sulfimide
, o-Sulfobenzimide
, o-Benzoic sulphimide
14900-0025
14900-1000
14900-5000
TS14900-0025EA
297
USD
TS14900-0025
TS14900-1000
TS14900-5000
Saccharin 98+%
Saccharin
Formula:
C₇H₅NO₃S MW: 183.19 g/mol Melting Pt: 226…230 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005866 CAS Number: 81-07-2 EINECS: 201-321-0 |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 226 to 230 °C |
Loss on drying | ≤1 % (105 °C, 2 hrs) |
Sulfated ash | ≤0.2 % |
Additional info | benzoate and salicylate: passes test |
Additional info | o-toluenesulfonamide: < 10 ppm |
Additional info | p-toluenesulfonamide: < 10 ppm |
Readily carbonizable substance | passes test |
Titration with NaOH | 98.0 to 101.0 % (On dried substance) |
Arsenic (As) | ≤3 ppm |
Selenium (Se) | ≤30 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...