Diphenylphosphinic acid 99%

Supplier: Thermo Scientific Chemicals

14871-0050 14871-0250
TS14871-0050EA 56.1 USD
TS14871-0050 TS14871-0250
Diphenylphosphinic acid 99%
Diphenylphosphinic acid
Formula: C₁₂H₁₁O₂P
MW: 218.19 g/mol
Melting Pt: 193…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00002132
CAS Number: 1707-03-5
EINECS: 216-948-5

Order Now

Specification Test Results

Appearance White Crystalline solid
Infrared spectrum Conforms
Melting point 193 to 196 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR