DL-Ethionine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-4-(ethylthio)butanoic acid

14617-0025 14617-0100 14617-0500
TS14617-0100EA 185 USD
TS14617-0100 TS14617-0025 TS14617-0500
DL-Ethionine 98%
DL-Ethionine
Formula: C₆H₁₃NO₂S
MW: 163.24 g/mol
Melting Pt: 265…268 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00063102
CAS Number: 67-21-0
EINECS: 200-647-0

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or flakes
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR