Norbornene 99% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Norbornylene

12924-0025 12924-5000 12924-0250
TS12924-0025EA 278.46 USD
TS12924-0025 TS12924-5000 TS12924-0250
Norbornene 99% stabilized
Norbornene
Formula: C₇H₁₀
Boiling Pt: 96 °C (760 mmHg)
Melting Pt: 42…46 °C
Flash Pt: –15 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00082304
CAS Number: 498-66-8
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Appearance White Crystalline mass
Infrared spectrum Conforms
Melting point 42 to 46 °C
GC ≥98.5 %
Stabilizer 0.1 % phenolic antioxidant
GC ≤0.05 % (diethylamine)
GC ≤0.05 % (ethanol)
Color scale ≤15 APHA
Residue after evaporation ≤0.002 %
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤10 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm
Heavy metals (as Pb) ≤1 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR