Formamidine acetate 99%

Supplier: Thermo Scientific Chemicals
Warning

11971-5000 11971-1000
TS11971-5000EA 189.72 USD
TS11971-5000 TS11971-1000
Formamidine acetate 99%
Formamidine acetate
Formula: C₃H₈N₂O₂
Melting Pt: 157…161 °C
Density: 1.124 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00012866
CAS Number: 3473-63-0
EINECS: 222-442-5

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR