Aminoiminomethanesulfinic acid 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Thiourea dioxideFormamidinesulfInic acid

11972-1000 11972-0100 11972-5000
TS11972-1000EA 29.27 USD
TS11972-1000 TS11972-5000 TS11972-0100
Aminoiminomethanesulfinic acid 99%
Aminoiminomethanesulfinic acid
Formula: CH₄N₂O₂S
Melting Pt: 126 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00002397
EINECS: 217-157-8
UN: 3341
ADR: 4.2,II

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤0.5 %
Titration with NaOH ≥98.5 %
Impurity ≤0.2 % Thiourea

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR