Ethoxymethylenemalononitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

11792-1000 11792-5000
TS11792-1000EA 113 USD
TS11792-1000 TS11792-5000
Ethoxymethylenemalononitrile 98%
Ethoxymethylenemalononitrile
Formula: C₆H₆N₂O
MW: 122.13 g/mol
Boiling Pt: 160 °C (12 mmHg)
Melting Pt: 63…69 °C
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00001854
CAS Number: 123-06-8
EINECS: 204-597-0
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow to orange-brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 63 to 68 °C
GC ≥97.5 %
Manganese (Mn) ≤2 ppm
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR