3-Chloropropyldimethylammonium chloride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Dimethylaminopropylchloride hydrochloride3-Dimethylaminopropyl chloride hydrochlorideN-(3-Chloropropyl)-N,N-dimethylammonium chloride3-Chloro-N,N-dimethylpropan-1-amine hydrochloride

11591-5000
TS11591-5000EA 128 USD
TS11591-5000
3-Chloropropyldimethylammonium chloride 99%
3-Chloropropyldimethylammonium chloride
Formula: C₅H₁₃Cl₂N
MW: 158.07 g/mol
Melting Pt: 139…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00012521
CAS Number: 5407-04-5
EINECS: 226-467-2

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (105 °C, 3 hrs)
Titration with HClO4 98.5 to 101.5 % (on dry substance)
Chloride (Cl) 21.9 to 22.9 % (titr.)
Chlorine (Cl) 44 to 45.5 % (total)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR