2-(Dimethylamino)ethanethiol hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Warning

11581-1000 11581-0050 11581-0250
TS11581-0050EA 61.1 USD
TS11581-0050 TS11581-0250 TS11581-1000
2-(Dimethylamino)ethanethiol hydrochloride 95%
2-(Dimethylamino)ethanethiol hydrochloride
Formula: C₄H₁₂ClNS
Melting Pt: 154…159 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012633
EINECS: 236-221-6

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration Iodimetric ≥94 %
GC ≥96.0 %
Enantiomeric excess ≥98.0 %
Specific optical rotation -7° to -9° (20 °C, 589 nm) (c=1, chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR