2-Chloroethyldiethylammonium chloride 99.5%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-(2-Chloroethyl)-N,N-diethylammonium chloride2-Diethylaminoethylchloride hydrochloride

11403-0010 11403-0025 11403-2500 11403-0050
TS11403-0025EA 213 USD
TS11403-0025 TS11403-2500 TS11403-0050 TS11403-0010
2-Chloroethyldiethylammonium chloride 99.5%
2-Chloroethyldiethylammonium chloride
Formula: C₆H₁₅Cl₂N
MW: 172.1 g/mol
Melting Pt: 208…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00012519
CAS Number: 869-24-9
EINECS: 212-786-4
UN: 2928
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 208 to 212 °C
Loss on drying ≤1 % (105 °C, 3 hrs)
Titration with HClO4 ≥99.4 % (On dry substance)
Zinc (Zn) ≤50 ppm
Heavy metals (as Pb) ≤30 ppm
Sugars, dextrin Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR