Diethyl acetamidomalonate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diethyl 2-acetamidomalonate

11393-0250 11393-5000 11393-1000
TS11393-1000EA 40.6 USD
TS11393-1000 TS11393-0250 TS11393-5000
Diethyl acetamidomalonate 99+%
Diethyl acetamidomalonate
Formula: C₉H₁₅NO₅
MW: 217.22 g/mol
Boiling Pt: 185 °C (20 mmHg)
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00009146
CAS Number: 1068-90-2
EINECS: 213-952-9

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 94 to 97 °C
GC ≥99.0 %
BSC Certification Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR