L(+)-Citrulline 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Cit-OH

11047-0250 11047-1000
TS11047-1000EA 299.88 USD
TS11047-1000 TS11047-0250
L(+)-Citrulline 99%
L(+)-Citrulline
Formula: C₆H₁₃N₃O₃
Melting Pt: 214 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064397
CAS Number: 372-75-8
EINECS: 206-759-6

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.3 % (1 g, 105 °C, 3 hrs)
Residue after ignition ≤0.1 %
UV transmittance ≥95.0 % (c=10, H2O, 430 nm) 1 cm cell
Specific optical rotation +24.5° to +26.8° (20 °C, 589 nm) (c=8, 6N HCl)
Titration with HClO4 98.5 to 101.0 %
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤200 ppm
Ammonium (NH4) ≤200 ppm
Arsenic (As) ≤1 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR