DL-Carnitine hydrochloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
10845-1000
10845-0250
TS10845-0250EA
33.46
USD
TS10845-0250
TS10845-1000
DL-Carnitine hydrochloride 99%
DL-Carnitine hydrochloride
Formula:
C₇H₁₆ClNO₃ Melting Pt: 190…205 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011904 CAS Number: 461-05-2 EINECS: 207-309-1 |
Specification Test Results
Appearance | White to off-white Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 194 to 200 °C |
Specific optical rotation | -1° to +1° (20 °C, 589 nm) (c=1, H2O) |
Titration with HClO4 | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...