4-Aminomorpholine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Aminomorpholine

10411-0250 10411-0050
TS10411-0050EA 51.41 USD
TS10411-0050 TS10411-0250
4-Aminomorpholine 95%
4-Aminomorpholine
Formula: C₄H₁₀N₂O
Density: 1.05 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00006174
CAS Number: 4319-49-7
EINECS: 224-347-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4770 to 1.4800 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR