Dodecylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dodecan-1-amine1-Aminododecane1-Dodecylamine

11766-1000 11766-0010 11766-5000
TS11766-5000EA 69 USD
TS11766-5000 TS11766-1000 TS11766-0010
Dodecylamine 98%
Dodecylamine
Formula: H₃C(CH₂)₁₀CH₂NH₂
MW: 185.35 g/mol
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 28…32 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008154
CAS Number: 124-22-1
EINECS: 204-690-6
UN: 3259
ADR: 8,II

Order Now

Specification Test Results

Appearance White Low melting crystalline solid
Infrared spectrum Conforms
Melting point 27 to 31 °C
GC ≥97.5 % (Alkyl chain distribution C12)
Titration with HClO4 ≥97.5 %
Amine number, total 297 to 304 mg KOH/g
Residue after evaporation ≤0.005 %
Titrable acid ≤0.002 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR