Cyclooctylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Aminocyclooctane

11146-0050 11146-1000 11146-0250
TS11146-1000EA 271.32 USD
TS11146-1000 TS11146-0050 TS11146-0250
Cyclooctylamine 97%
Cyclooctylamine
Formula: C₈H₁₇N
MW: 127.23 g/mol
Boiling Pt: 190 °C (760 mmHg)
Melting Pt: –48 °C
Density: 0.92 g/cm³
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00001748
CAS Number: 5452-37-9
EINECS: 226-694-7
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4800 to 1.4820 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR