1-Aminocyclopentanecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cycloleucine1-Aminocyclopentane-1-carboxylic acid

10347-0100 10347-0050
TS10347-0100EA 182.58 USD
TS10347-0100 TS10347-0050
1-Aminocyclopentanecarboxylic acid 97%
1-Aminocyclopentanecarboxylic acid
Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 320…322 °C
Storage Temperature: Ambient
MDL Number: MFCD00001381
CAS Number: 52-52-8
EINECS: 200-144-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or flakes
Infrared spectrum Conforms
Titration with HClO4 ≥96.0 %
GC ≥96.0 %
Specific optical rotation +42° to +48° (20°C, 589 nm) (c=3, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR