Chlorhexidine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,1'-Hexamethylenebis[5-(p-chlorophenyl)biguanide]

46102-0050 46102-0010
TS46102-0050EA 342.02 USD
TS46102-0050 TS46102-0010
Chlorhexidine 98%
Chlorhexidine
Formula: C₂₂H₃₀Cl₂N₁₀
MW: 505.4466 g/mol
Melting Pt: 134…136 °C
Storage Temperature: Ambient
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 130 to 136 °C
Water ≤1 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR