(±)-1-(1-Naphthyl)ethylamine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(Naphthalen-1-yl)ethanamine

45057-0050 45057-1000
TS45057-0050EA 42.02 USD
TS45057-0050 TS45057-1000
(±)-1-(1-Naphthyl)ethylamine 98%
(±)-1-(1-Naphthyl)ethylamine
Formula: C₁₂H₁₃N
MW: 171.24 g/mol
Boiling Pt: 156 °C (15 mmHg)
Density: 1.063 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004014
CAS Number: 42882-31-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6220 to 1.6240 (20°C, 589 nm)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR