4-(N,N-Dimethylamino)phenylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N,N-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline4-(Dimethylamino)phenylboronic acid pinacol ester

44496-0010 44496-0050
TS44496-0010EA 81 USD
TS44496-0010 TS44496-0050
4-(N,N-Dimethylamino)phenylboronic acid pinacol ester 97%
4-(N,N-Dimethylamino)phenylboronic acid pinacol ester
Formula: C₁₄H₂₂BNO₂
MW: 247.1410 g/mol
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD05663854
CAS Number: 171364-78-6

Order Now

Specification Test Results

Appearance Off-white to light green to tan Powder or solid
Infrared spectrum Conforms
Melting point 122 to 127 °C
GC ≥96 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR