1-Methyl-4-piperidinemethanol 97%

Supplier: Thermo Scientific Chemicals
Warning

44048-0010 44048-0050
TS44048-0010EA 52.12 USD
TS44048-0010 TS44048-0050
1-Methyl-4-piperidinemethanol 97%
1-Methyl-4-piperidinemethanol
Formula: C₇H₁₅NO
Melting Pt: 29…31 °C
Density: 0.98 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD03411903
CAS Number: 20691-89-8

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
Titration with HCl ≥96.0 to ≤104.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR