N-Phenyl-2-naphthylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

43991-0250 43991-1000
TS43991-0250EA 50.69 USD
TS43991-0250 TS43991-1000
N-Phenyl-2-naphthylamine 97%
N-Phenyl-2-naphthylamine
Formula: C₁₆H₁₃N
Boiling Pt: 395 °C
Melting Pt: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00004052
CAS Number: 135-88-6
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Off-white to grey to brown Crystalline powder
Infrared spectrum Conforms
Melting point 105 to 108 °C
HPLC ≥96.0 %
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR