N-tert-Butylcyclohexylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

43983-0050 43983-0010
TS43983-0010EA 70 USD
TS43983-0010 TS43983-0050
N-tert-Butylcyclohexylamine 97%
N-tert-Butylcyclohexylamine
Formula: C₁₀H₂₁N
Boiling Pt: 172…174 °C (760 mmHg)
Density: 0.834 g/cm³
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00075598
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR