1-Methylindol-3-ylacetic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Methyl-3-indoleacetic acid2-(1-Methyl-1H-indol-3-yl)acetic acid

43924-0010 43924-0050
TS43924-0010EA 35.9 USD
TS43924-0010 TS43924-0050
1-Methylindol-3-ylacetic acid 98%
1-Methylindol-3-ylacetic acid
Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
Melting Pt: 125…126.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00130160
CAS Number: 1912-48-7

Order Now

Specification Test Results

Appearance White to tan Crystalline powder
Infrared spectrum Conforms
Melting point 122 to 128 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR