1H-Indazole-6-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

43012-0010 43012-0050
TS43012-0010EA 73.03 USD
TS43012-0010 TS43012-0050
1H-Indazole-6-carboxylic acid 97%
1H-Indazole-6-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.1454 g/mol
Melting Pt: 297…300 °C
Storage Temperature: Ambient
MDL Number: MFCD06804571
CAS Number: 704-91-6

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Titration with HClO4 ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR