Methyl-2-aminoisonicotinate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-aminopyridine-4-carboxylateMethyl 2-aminoisonicotinate

43006-0010 43006-0050
TS43006-0010EA 59.26 USD
TS43006-0010 TS43006-0050
Methyl-2-aminoisonicotinate 98%
Methyl 2-aminoisonicotinate
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Melting Pt: 145…148 °C
Storage Temperature: Ambient
MDL Number: MFCD04039316
CAS Number: 6937-03-7

Order Now

Specification Test Results

Appearance White to brown Crystalline powder
Infrared spectrum Conforms
Melting point 144 to 148 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR