1H-Indazole-5-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

42956-0010 42956-0050
TS42956-0010EA 81.29 USD
TS42956-0010 TS42956-0050
1H-Indazole-5-carboxylic acid 97%
1H-Indazole-5-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 321…324 °C
Storage Temperature: Ambient
MDL Number: MFCD06804570
CAS Number: 61700-61-6

Order Now

Specification Test Results

Appearance White to brown Crystalline powder or powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR