2-Amino-4-thiazolecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Aminothiazole-4-carboxylic acid

42948-0010 42948-0050
TS42948-0010EA 79.66 USD
TS42948-0010 TS42948-0050
2-Amino-4-thiazolecarboxylic acid 97%
2-Amino-4-thiazolecarboxylic acid
Formula: C₄H₄N₂O₂S
MW: 144.15 g/mol
Melting Pt: 238…239 °C
Storage Temperature: Ambient
MDL Number: MFCD00859429
CAS Number: 40283-41-8

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Water ≤0.1 % (Coulometric)
GC ≥99.0 %
GC ≤0.1 % 2-Propanol
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm
Acidity (CH3COOH) ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR