Isoindoline 97%

Supplier: Thermo Scientific Chemicals
Warning

42628-0050 42628-0010
TS42628-0010EA 115 USD
TS42628-0010 TS42628-0050
Isoindoline 97%
Isoindoline
Formula: C₈H₉N
Boiling Pt: 221 °C (760 mmHg)
Melting Pt: 17 °C
Density: 1.05 g/cm³
Flash Pt: 94 °C
Storage Temperature: Freezer
MDL Number: MFCD00605324
CAS Number: 496-12-8

Order Now

Specification Test Results

Appearance Clear colorless to yellow to pink to orange-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5670 to 1.5710 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR