Xylene cyanole, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: C.I. 43535Cyanol FF

42269-5000 42269-1000 42269-0050 42269-0500 42269-0250
TS42269-0500EA 286 USD
TS42269-0500 TS42269-0050 TS42269-5000 TS42269-0250 TS42269-1000
Xylene cyanole, pure
Xylene cyanole
Formula: C₂₅H₂₇N₂NaO₇S₂
MW: 554.61 g/mol
MDL Number: MFCD00019481
CAS Number: 4463-44-9
EINECS: 224-728-5

Order Now

Specification Test Results

Appearance Dark red to dark blue Crystalline powder
Infrared spectrum Conforms
Dye content ≥80 %
GC ≥99.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR