N-Benzylaniline 99%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Phenylbenzylamine((Phenylmethyl)amino)benzene

41739-0250 41739-1000 41739-0050
TS41739-0050EA 28.25 USD
TS41739-0050 TS41739-1000 TS41739-0250
N-Benzylaniline 99%
N-Benzylaniline
Formula: C₆H₅CH₂NHC₆H₅
MW: 183.25 g/mol
Boiling Pt: 306…307 °C (760 mmHg)
Melting Pt: 34…38 °C
Density: 1.06 g/cm³
Flash Pt: 152 °C
Storage Temperature: Ambient
MDL Number: MFCD00003018
CAS Number: 103-32-2
EINECS: 203-100-4

Order Now

Specification Test Results

Appearance Colorless to yellow to light beige Crystalline powder or low melting mass
Infrared spectrum Conforms
Melting point 34 to 38 °C
GC ≥98.5 %
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR