Methyl-1,2,4-triazole-3-carboxylate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 1H-1,2,4-triazole-3-carboxylate

41500-0250 41500-0050
TS41500-0250EA 135.66 USD
TS41500-0250 TS41500-0050
Methyl-1,2,4-triazole-3-carboxylate 98%
Methyl 1,2,4-triazole-3-carboxylate
Formula: C₄H₅N₃O₂
MW: 127.10 g/mol
Melting Pt: 190…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00135989
CAS Number: 4928-88-5

Order Now

Specification Test Results

Appearance White Powder and/or flakes
Infrared spectrum Conforms
Melting point 185 to 193 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR