1,1-Dimethylguanidinium sulfate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,1-Dimethylguanidine Sulfate(2:1)N,N-Dimethylguanidine sulphateN,N-Dimethylguanidine sulfate

40835-0100 40835-0500 40835-1000
TS40835-0500EA 189.72 USD
TS40835-0500 TS40835-1000 TS40835-0100
1,1-Dimethylguanidinium sulfate 98%
1,1-Dimethylguanidinium sulphate
Formula: C₆H₂₀N₆O₄S
MW: 272.33 g/mol
Melting Pt: 280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013131
CAS Number: 598-65-2
EINECS: 209-946-0

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR